Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Trypticase™ Soy Agar w/5% Sheep Blood/ Chocolate II, I Plate™ Prepared Media
Two media formulations in single plate for cultivating fastidious microorganisms
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Color | Dark Brown |
Form | Liquid |
Manufacturing Origin | United States |
Formulation | Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, Defibrinated Sheep Blood 5%, Pancreatic digest of Casein 7.5g/L, Selected meat Peptone 7.5g/L, Corn Starch 1g/L, Dipotassium |
Catalog Number | Mfr. No. | For Use With (Application) | dimMediaType | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | dimMediaType | Quantity | Price | Quantity & Availability | |||
L21303
|
BD
221303 |
Cultivation of fastidious microorganisms and for visualization of hemolytic reaction | 100/Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Tripticase Soy Agar (TSA II)
- For the visualization of hemolytic reactions produced by many bacterial species
- GC II Agar with Hemoglobin and IsoVitaleX™
- For use in qualitative procedures for isolation and cultivation of fastidious microorganisms, Neisseria and Haemophilus species
Specifications
ISO/DIS, 11014 | |
Liquid | |
Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, Defibrinated Sheep Blood 5%, Pancreatic digest of Casein 7.5g/L, Selected meat Peptone 7.5g/L, Corn Starch 1g/L, Dipotassium |
Dark Brown | |
United States | |
Plate |
Safety and Handling
ShelfLife : 84 Days
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title