Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Bcl-2 Related Sampler Kit

For blotting procedures

Manufacturer:  BD 612742

Catalog No. BDB612742


Add to cart

Alert:

Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing


Includes: Bad, BAG-1, Bax, Bcl-2, Bcl-x, Bcl-x, Beclin, Bid, Storage Buffer: aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide

Description

Description

  • Concentration: 250µg/mL
  • Format: Purified

Western Blotting

Specifications

Specifications

1 Kit
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research use only