Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Caspase-3, Active Form, mAb Apoptosis Kit

Specific recognision of active form of caspase-3 in human and mouse cells

Manufacturer:  BD 550914

Catalog No. BDB550914


Add to cart

Includes: PE-conjugated Monoclonal Rabbit Anti-Active Caspase-3 Antibody, BD Cytofix/Cytoperm™ Fixation/Permeabilization Solution, and BD Perm/Wash™ Buffer

Description

Description

Cysteine proteases plays key role in apoptosis and inflammation. Caspase-3 being key protease is activated during early stages of apoptosis and, like other members of caspase family, is synthesized as inactive pro-enzyme that is processed in cells undergoing apoptosis by self-proteolysis and/or cleavage by another protease. The processed forms of caspases consist of large (17-22 kDa) and small (10-12 kDa) subunits which associate to form active enzyme. Active caspase-3, a marker for cells undergoing apoptosis, consists of heterodimer of 17 and 12 kDa subunits which is derived from the 32 kDa pro-enzyme. Active caspase-3 proteolytically cleaves and activates other caspases, as well as relevant targets in cytoplasm and nucleus. This antibody has been reported to specifically recognize the active form of caspase-3 in human and mouse cells.

  • Clone: C92-605
  • Format: PE
  • Isotype: IgG

Intracellular staining (flow cytometry)

Specifications

Specifications

1 Kit
Liquid
Intracellular staining
4°C
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research use only