Learn More
BD Caveolin Sampler Kit
Manufacturer:  BD 611766
Catalog No. BDB611766
Add to cart
Includes: Caveolin 1, Caveolin (pY14), Caveolin 2, Caveolin 3, Flotillin-1, Flotillin-2/ESA, GPI-Phospholipase D, eNOS/NOS Type III, nNOS/NOS Type I
Description
Description
  • Storage Buffer: Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide
Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry
Specifications
Specifications
250μg/mL
1 Kit