Learn More
Gibco™ Cell MAb Medium, animal component free
Produces yields higher than classical basal media with similar supplementation
$93.60
Specifications
Quantity 1000mL
Storage 4°C
View More Specs
Products 1
Catalog Number Mfr. No. Without Additives Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Without Additives Product Type Price Quantity & Availability  
CB220513
View Documents
Gibco™
220513
Phenol Red or Pluronic™ Acid or Other Surfactants Cell MAb Medium, animal component free Each for $93.60
Add to cart
Description
Description
Promotes an acceleration in the monoclonal antibody secretion rates of cells in a batch mode for performance in hollow fiber and stir tank systems.
  • Suitable for use in perfusion systems
  • Shown to increase purified monoclonal antibody yields by 5 to 25 times over classical basal media with similar supplementation
  • Reduce media consumption and labor cost, reduce requirements for media storage and preparation time, decrease labor required to perform continuous feeding and decrease contamination because of decreased handling of the culture
  • Concentration step is eliminated in most cases, and final bulk harvest is reduced, producing less material to run through the purification process
  • Only minimal adaptation is needed
  • Hybridomas grown in MAb Basal Medium can be supplemented in the same way as those grown in traditional basal media
  • MAb Serum-Free and Animal-Free are complete media and do not require serum supplementation; Basal Medium does require serum supplementation
Specifications
Specifications
1000mL
4°C