Learn More
BD Clay Adams Fixed-Angle Rotors for Dynac and Dynac III Centrifuges
For use with Dynac and Dynac III Centrifuges
$1614.00
Specifications
For Use With (Equipment) | Dynac™ and Dynac™ III Centrifuge |
---|---|
Type | Fixed Angle Rotor |
Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
05-100-34
|
BD
420114 |
24 x 15mL |
Each for $1,614.00
|
|
Description
- Six-, 12-, and 24-place fixed-angle rotors accept tubes from 3 to 15mL when equipped with buckets (sold separately) For optimum performance, the use of heavy-duty glassware is recommended in these rotors
Specifications
Dynac™ and Dynac™ III Centrifuge | |
Fixed Angle Rotor |