Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Coated Vesicle Sampler Kit

For routine western blotting analysis

Manufacturer:  BD 611424

Catalog No. BDB611424


Add to cart

Includes: Adaptin a, Adaptin b, Adaptin g, Adaptin d, Amphiphysin, AP180, Clathrin Heavy Chain, EEA1, eps15, b-NAP, p47A, Rab4

Description

Description

  • Format: Purified
  • Storage buffer: Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide

Western Blotting

Specifications

Specifications

1 Kit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only