Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Cytometric Bead Array (CBA) Mouse IFN-γ Enhanced Sensitivity Flex Set ,Bead A4
Sufficient for 100 tests
Supplier: BD 562233
Includes: Mouse IFN-γ Standard, Mouse IFN-γ Enhanced Sensitivity Capture Bead A4, Mouse IFN-γ Detection Reagent (Part A)
Description
Host Species:
Species Reactivity: Mouse
Flow Cytometry
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction