Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Cytometric Bead Array Human IL-8 Flex Set Standard
Used as standard control
$90.00 - $100.50
Specifications
Content And Storage | 4°C |
---|---|
Format | Lyophilized |
Includes | Lyophilized recombinant Human Interferon-l (IFN-l) |
Quantity | 0.2 mL |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
BDB558456
|
BD
558456 |
Human IFN-gamma Flex Set Standard |
Each for $100.50
|
|
BDB558457
|
BD
558457 |
Human IL-1beta Flex Set Standard |
Each for $100.50
|
|
BDB558464
|
BD
558464 |
Human IL-6 Flex Set Standard |
Each for $90.00
|
|
BDB558462
|
BD
558462 |
Human IL-4 Flex Set Standard |
Each for $100.50
|
|
BDB558461
|
BD
558461 |
Human IL-2 Flex Set Standard |
Each for $90.00
|
|
BDB558458
|
BD
558458 |
Human IL-10 Flex Set Standard |
Each for $90.00
|
|
BDB558472
|
BD
558472 |
Human TNF Flex Set Standard |
Each for $90.00
|
|
BDB558465
|
BD
558465 |
Human IL-8 Flex Set Standard |
Each for $100.50
|
|
BDB558474
|
BD
558474 |
Mouse IL-10 Flex Set Standard |
Each for $90.00
|
|
BDB558479
|
BD
558479 |
Mouse IL-6 Flex Set Standard |
Each for $90.00
|
|
BDB558480
|
BD
558480 |
Mouse TNF Flex Set Standard |
Each for $100.50
|
|
Description
This product contains lyophilized recombinant human Interferon-λ (IFN-λ)
- Storage Buffer: Lyophilized in aqueous buffered solution containing BSA and ProClin™ 150
Flow cytometry
Specifications
4°C | |
Lyophilized recombinant Human Interferon-l (IFN-l) | |
RUO |
Lyophilized | |
0.2 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title