Learn More
BD Difco™ Dehydrated Media Tryptic Soy Broth, Select APS
Used in qualitative procedures for cultivation of fastidious and nonfastidious microorganisms from variety of clinical and nonclinical specimens
Manufacturer:  BD 214887
Catalog No. 22-350-781
Add to cart
Description
Description
  • Select Alternative Protein Source (APS)
  • Obtained from animal-free components
Specifications
Specifications
Liquid
Pail
10,000g
Tryptic Soy broth