Learn More
BD BBL™ Plated Media CHROMagar™ Listeria (AOAC™-RI Approved)
Selective medium for the isolation, differentiation and identification of Listeria monocytogenes and L. ivanovii from food and environmental samples
Manufacturer:  BD 215085
Catalog No. 14-432-40
Add to cart
This item is not returnable. View return policy
Description
Description
  • Faster time to results
  • Higher accuracy, sensitivity and specificity in food matrix testing
  • Detection level: 1 to 18 CFU/25g
  • Colonies revealed in distinctive, identifiable colors
  • Greater recovery from raw ground beef compared to USDA methods
Certifications
AOAC™ Research Institute (AOAC-RI) under the Performance Tested Methods Program using FDA BAM, USDA FSIS, and ISO methods
Specifications
Specifications
20/Pk.