Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Listeria O Antisera
Used as positive control in identification of Listeria monocytogenes in the slide agglutination test
Supplier: BD 223021
This item is not returnable.
View return policy
Description
Listeria monocytogenes is a widespread problem in public health and the food industries.
- Lyophilized, polyclonal rabbit antisera containing approximately 0.1% sodium azide as a preservative
- Listeria O Antisera Types 1 and 4 are specific for the respective serotypes of L. monocytogenes while Listeria O Antiserum Poly contains agglutinins for L. monocytogenes serotypes 1 and 4
For in vitro Diagnostic Use.
Certifications
CE marked
Specifications
CE Marked; For in vitro diagnostic use | |
Antiserum | |
1 mL |
Refrigerate, store at 2°C to 8°C | |
Used as positive control in identification of Listeria monocytogenes in the slide agglutination test |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction