Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Listeria O Antisera

Used as positive control in identification of Listeria monocytogenes in the slide agglutination test

$189.00 - $196.00

Specifications

Certifications/Compliance CE Marked; For in vitro diagnostic use
For Use With (Application) Used as positive control in identification of Listeria monocytogenes in the slide agglutination test
Quantity 1mL
Storage Requirements Refrigerate, store at 2° to 8°C
View More Specs

Products 3
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
DF2300-50-2
View Documents
BD
223001
Listeria O Antiserum Type 1
Each for $196.00
Add to cart
 
DF2301-50-1
View Documents
BD
223011
Listeria O Antiserum Type 4
Each for $194.00
Add to cart
 
DF2302-50-0
View Documents
BD
223021
Listeria O Antiserum Poly Types 1 and 4
Each for $189.00
Add to cart
 
Description

Description

Listeria monocytogenes is a widespread problem in public health and the food industries.

  • Lyophilized, polyclonal rabbit antisera containing approximately 0.1% sodium azide as a preservative
  • Listeria O Antisera Types 1 and 4 are specific for the respective serotypes of L. monocytogenes while Listeria O Antiserum Poly contains agglutinins for L. monocytogenes serotypes 1 and 4

For in vitro Diagnostic Use.

Certifications

CE marked

Specifications

Specifications

CE Marked; For in vitro diagnostic use
Used as positive control in identification of Listeria monocytogenes in the slide agglutination test
1mL
Refrigerate, store at 2° to 8°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit