Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Shigella Poly Antisera

Used in serologic identification of Shigella

Manufacturer:  BD 228381

 View more versions of this product

Catalog No. DF2838-47-9


Due to product restrictions, please to purchase or view availability for this product.

This item is not returnable. View return policy

Description

Description

  • Available in 3.0mL packaged vials

Specifications

Specifications

2° to 8°C
Used for serologic identification of Shigella
Liquid
Alkalescens Dispar Antiserum poly
3mL
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit