Learn More
BD Falcon™ Roller Bottles, Vented Cap
Compatible with manual and automated filling systems
Manufacturer:  BD 353069
 View more versions of this product
Catalog No. 08-772H
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Ideal for use in research and manufacturing applications involving the scale-up of mammalian cells for purposes of virus propagation and bioproduct production.
  • Available with smooth 850cm2 growth area configuration or pleated 1450cm2 growth area configuration
  • One-piece BD Tufrol™ polystyrene manufactured using a proprietary process that provides high impact resistance
  • Sterilized by gamma irradiation, nonpyrogenic
  • Vented cap has integral hydrophobic 0.2µm membrane filter
Custom packaging and lot control available. Contact your Fisher Scientific Customer Service Representative.
Specifications
Specifications
Pleated
Polystyrene
1450cm2