Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD General Use and PrecisionGlide Hypodermic Needles

Fit any Luer-Lok™, slip tip, or eccentric tip syringe

Manufacturer:  BD 305195

 View more versions of this product

Catalog No. 14-826-5G



 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Specifications

Siliconized
0.048 in.
Yes
18g
Protective Plastic sheath
1 in.
Bevel point
Stainless Steel
Yes
Luer-Lok
0.124cm
Any Luer-Lok, slip tip, or eccentric tip syringe
Polypropylene
Yes
2.5cm
Regular
Precisionglide™ Hypodermic Conventional Needle
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit