Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD HCT-8 Cell Lysate

Convenient to use and provided in denatured format allowing for immediate loading onto an SDS-PAGE gel

Manufacturer:  BD 611474

Catalog No. BDB611474


Add to cart

Includes: Human Ileocecal Adenocarcinoma

Description

Description

  • Rich in endogenously expressed proteins
  • Useful control for western blotting application
  • Concentration: 1.0mg/mL
  • Storage Buffer: SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)

Specifications

Specifications

Ileocecal Adenocarcinoma
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
Human
Western Blot
500μg
-20°C
1mg/mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit