Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD HeLa + Anisomycin Lysate

Convenient to use and provided in denatured format allowing for immediate loading onto SDS-PAGE gel

Manufacturer:  BD 611692

Catalog No. BDB611692


Add to cart

Includes: Human Epitheloid Cervical Carcinoma

Description

Description

  • Useful control for western blotting application
  • Source that can be rich in endogenously expressed proteins
  • Storage Buffer: SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)

Specifications

Specifications

Epitheloid Cervical Carcinoma
500μg
-20°C
1mg/mL
HeLa (Human Epitheloid Carcinoma) cells, were grown to 80 to 90% confluency, then treated with 25μg/mL Anisomycin, an antibiotic and Protein synthesis inhibitor, for 15 min. at 37°C
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
Human
Western Blot
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit