Learn More
BD HeLa Cell Lysate
Cell or tissue lysates are a useful control for the western blotting application
Manufacturer:  BD 611449
Catalog No. BDB611449
Add to cart
Includes: Human Epitheloid Cervical Carcinoma
Description
Description
A source that can be rich in endogenously expressed proteins, this lysate is convenient to use and provided in a denatured format allowing for immediate loading onto an SDS-PAGE gel.
  • Storage buffer: 1mg/mL in SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)
Western blot
Specifications
Specifications
Western Blot
500 μg
Human
Epitheloid Cervical Carcinoma
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
1 mg/mL
Safety and Handling
Safety and Handling
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Research Use Only