Learn More
BD HeLa + Pervanadate Lysate
Useful control for the western blotting application
Manufacturer:  BD 612015
Catalog No. BDB612015
Add to cart
Includes: Human Epitheloid Cervical Carcinoma
Description
Description
  • Rich in endogenously expressed proteins
  • Convenient to use and provided in a denatured format allowing for immediate loading onto an SDS-PAGE gel
Specifications
Specifications
Western Blot
HeLa (Human Epitheloid Carcinoma) cells, were grown to 80 to 90% confluency, then treated with 1mM pervanadate, a general inhibitor of Protein Tyrosine Phosphatases, for 15 min. at 37°C
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
1 mg/mL
Epitheloid Cervical Carcinoma
500 μg
Human
Safety and Handling
Safety and Handling
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Research Use Only