Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Mouse Anti-Human CD127

CD127 is expressed on thymocytes, T- and B-cell progenitors, mature T cells, and some lymphoid and myeloid cells. BD Horizon™ BV480 Mouse Anti-Human CD127 is down-regulated following T cell activation. Studies indicate that the IL-7 Receptor plays an important role in the proliferation and differentiation of mature T cells.

Manufacturer:  BD 566101

 View more versions of this product

Catalog No. BDB566101


Add to cart

Description

Description

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon BV480 can be excited by the violet laser and detected in the BD Horizon BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications

Specifications

100 Tests
Flow cytometry
3575
5 μL
Store undiluted at 4°C
QC Testing: Human
BV480
BV480 Mouse Anti-Human CD127
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit