Learn More
BD BV480 Mouse Anti-Human CD20
CD20 is expressed on pre-B-cells, resting and activated B cells, and follicular dendritic cells, but not plasma cells. BD Horizon™ BV480 Mouse Anti-Human CD20 is observed in low levels on a small subset of normal circulating T lymphocytes. The CD20 molecule is involved in the regulation of B-cell activation.
Supplier: BD 566132
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Human CD20 | |
Flow Cytometry | |
BV480 | |
Store undiluted at 4°C | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
100 Tests | |
QC Testing: Human; Tested in Development: Rhesus, Cynomolgus, Baboon | |
5 μL | |
931 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.