Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Mouse Anti-Human CD28

The CD28.2 monoclonal antibody specifically binds to CD28, a 44kDa homodimeric transmembrane glycoprotein present on most mature T cells, thymocytes and plasma cells. BD Horizon™ BV480 Mouse Anti-Human CD28 is a costimulatory receptor that binds CD80 and CD86 as ligands and plays a very important role in T cell-B cell interactions.

$175.00 - $386.00

Specifications

For Use With (Application) Flow cytometry
Cross Reactivity QC Testing: Human
Format BV480
Product Type BV480 Mouse Anti-Human CD28
Gene ID (Entrez) 940
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566173
View Documents
BD
566173
25 Tests
Each for $175.00
Add to cart
 
BDB566110
View Documents
BD
566110
100 Tests
Each for $386.00
Add to cart
 
Description

Description

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications

Specifications

Flow cytometry
BV480
940
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
QC Testing: Human
BV480 Mouse Anti-Human CD28
5 μL
Store undiluted at 4°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit