Learn More
BD BV480 Mouse Anti-Human CD49d
The 9F10 monoclonal antibody specifically reacts with the integrin α4 chain, that is expressed as a heterodimer with either of two β integrin subunits, β 1 (CD29) or β7. BD Horizon™ BV480 Mouse Anti-Human CD49d clone cross-reacts with a subset of peripheral blood lymphocytes, monocytes, and some granulocytes of baboon and both rhesus and cynomolgus macaque monkeys.
Manufacturer:  BD 566183
 View more versions of this product
Catalog No. BDB566183
Add to cart
Description
Description
  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
Specifications
QC Testing: Human; Tested in Development: Rhesus, Cynomolgus, Baboon, Dog, Cow, Sheep, Cat, Horse
BV480
25 Tests
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Flow cytometry
BV480 Mouse Anti-Human CD49d
5 μL
Store undiluted at 4°C