Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BV480 Mouse Anti-Human CD49d
The 9F10 monoclonal antibody specifically reacts with the integrin α4 chain, that is expressed as a heterodimer with either of two β integrin subunits, β 1 (CD29) or β7. BD Horizon™ BV480 Mouse Anti-Human CD49d clone cross-reacts with a subset of peripheral blood lymphocytes, monocytes, and some granulocytes of baboon and both rhesus and cynomolgus macaque monkeys.
Specifications
Product Type | BV480 Mouse Anti-Human CD49d |
---|---|
For Use With (Application) | Flow Cytometry |
Cross Reactivity | QC Testing: Human; Tested in Development: Rhesus, Cynomolgus, Baboon, Dog, Cow, Sheep, Cat, Horse |
Form | BV480 |
Sample Volume | 5 μL |
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Human CD49d | |
QC Testing: Human; Tested in Development: Rhesus, Cynomolgus, Baboon, Dog, Cow, Sheep, Cat, Horse | |
5 μL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Flow Cytometry | |
BV480 | |
Store undiluted at 4°C |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title