Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Mouse Anti-Human CD62L

CD62L is expressed on neutrophils, monocytes, T- and B-lymphocyte subsets and NK cells. BD Horizon™ BV480 Mouse Anti-Human CD62L DREG-56 antibody recognizes the same antigen as LAM-1, and specifically inhibits >90% of binding of human lymphocytes to high endothelial venules (HEV) in frozen

sections of peripheral, but not mucosal lymphoid tissue.

$185.00 - $396.00

Specifications

For Use With (Application) Flow cytometry
Cross Reactivity QC Testing: Human
Format BV480
Product Type BV480 Mouse Anti-Human CD62L
Sample Volume 5 μL
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566174
View Documents Promotion Details
BD
566174
25 Tests
Each for $185.00
Add to cart
 
BDB566111
View Documents Promotion Details
BD
566111
100 Tests
Each for $396.00
Add to cart
 
Description

Description

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications

Specifications

Flow cytometry
BV480
5 μL
Store undiluted at 4°C
QC Testing: Human
BV480 Mouse Anti-Human CD62L
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit