Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Mouse Anti-Human CD7

CD7 is expressed on thymocytes, T cells, pre-B cells, and NK cells. BD Horizon™ BV480 Mouse Anti-Human CD7 is present in reduced density on monocytic cells and cell lines. Functional studies demonstrate that crosslinking of the CD7 can induce transmembrane calcium flux in T cells.

Manufacturer:  BD 566161

 View more versions of this product

Catalog No. BDB566161


Add to cart

Description

Description

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications

Specifications

25 Tests
Flow cytometry
924
5 μL
Store undiluted at 4°C
QC Testing: Human
BV480
BV480 Mouse Anti-Human CD7
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit