Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Mouse Anti-Human CD86

CD86 (B70/B7-2) is expressed primarily on monocytes, dendritic cells and activated B cells. BD Horizon™ BV480 Mouse Anti-Human CD86 is a ligand for CD28 and CTLA-4 and plays an important role in costimulation of T cells in primary immune response.

$185.00 - $380.00

Specifications

For Use With (Application) Flow cytometry
Cross Reactivity QC Testing: Human; Tested in Development: Rhesus, Cynomolgus, Baboon
Format BV480
Product Type BV480 Mouse Anti-Human CD86
Sample Volume 5 μL
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566180
View Documents
BD
566180
25 Tests
Each for $185.00
Add to cart
 
BDB566131
View Documents
BD
566131
100 Tests
Each for $380.00
Add to cart
 
Description

Description

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications

Specifications

Flow cytometry
BV480
5 μL
Store undiluted at 4°C
QC Testing: Human; Tested in Development: Rhesus, Cynomolgus, Baboon
BV480 Mouse Anti-Human CD86
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit