Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Mouse Anti-Human IgM

IgM is an important component in the first line of defense against foreign pathogens, but may also play a role in autoimmune diseases. BD Horizon™ BV480 Mouse Anti-Human IgM monomers consist of two light and two heavy chains. Unlike the heavy chain of an IgG antibody which contains 3 constant Ig domains, the μ heavy chain of IgM contains 4 constant Ig domains.

Manufacturer:  BD 566146

 View more versions of this product

Catalog No. BDB566146


Add to cart

Description

Description

With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications

Specifications

100 Tests
Flow cytometry
BV480 Mouse Anti-Human IgM
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
QC Testing: Human
BV480
5 μL
Store undiluted at 4°C
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit