Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BV480 Mouse Anti-Human TCR γδ
The γδ TCR is associated with the signal-transducing CD3 complex. BD Horizon™ BV480 Mouse Anti-Human TCR γδ is expressed by thymocytes and by peripheral T cell subsets (γδ T cells) that are located in the blood, liver, skin and various lymphoid and mucosal tissues. γδ T cells contribute to both innate and adaptive immune responses to infections and tumors.
$214.00 - $462.00
Specifications
Product Type | BV480 Mouse Anti-Human TCR γδ |
---|---|
For Use With (Application) | Flow Cytometry |
Concentration | 0.2 mg/mL |
Cross Reactivity | QC Testing: Human |
Form | BV480 |
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon BV480 can be excited by the violet laser and detected in the BD Horizon BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Human TCR γδ | |
0.2 mg/mL | |
BV480 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Flow Cytometry | |
QC Testing: Human | |
Store undiluted at 4°C |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title