Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Mouse Anti-Mouse CD45.2

The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. BD horizon™ BV480 Mouse Anti-Mouse CD45.2 has 104 monoclonal antibody that recognizes the CD45 (Leukocyte Common Antigen) present on all leucocytes of most mouse strains.

Manufacturer:  BD 566073

 View more versions of this product

Catalog No. BDB566073


Add to cart

Description

Description

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications

Specifications

0.2 mg/mL
Flow cytometry
19264
0.1 mg
Store undiluted at 4°C
QC Testing: Mouse
BV480
BV480 Mouse Anti-Mouse CD45.2
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit