Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Mouse Anti-NHP CD45

D058-1283 is a CD45 monoclonal antibody specific for non-human primate leucocytes. BD Horizon™ BV480 Mouse Anti-NHP CD45 antibody reacts with baboon, Rhesus and Cynomolgus Macaque leucocytes in a similar pattern to CD45 binding to leukocyte common antigen (LCA) on human cells.  

$185.00 - $396.00

Specifications

For Use With (Application) Flow cytometry
Cross Reactivity QC Testing: Rhesus, Cynomolgus, Baboon
Sample Volume 5 μL
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Storage Requirements Store undiluted at 4°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566145
View Documents Promotion Details
BD
566145
100 Tests
Each for $396.00
Add to cart
 
BDB566152
View Documents Promotion Details
BD
566152
25 Tests
Each for $185.00
Add to cart
 
Description

Description

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications

Specifications

Flow cytometry
5 μL
Store undiluted at 4°C
BV480 Mouse Anti-NHP CD45
QC Testing: Rhesus, Cynomolgus, Baboon
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
BV480
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit