Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

BD BV480 Rat Anti-Mouse CD117

Widespread expression of CD117 in hematopoietic precursors is consistent with participation of c-Kit and its ligand in regulation of several hematopoietic lineages. BD Horizon™ BV480 Rat Anti-Mouse CD117 is also involved in the regulation of some events during the development of T lymphocytes. CD117 is also expressed by mast cells and by dendritic cells found in the periarteriolar lymphocytoc sheaths (T-cell areas) of splenic white pulp.

Specifications

For Use With (Application) Flow cytometry
Concentration 0.2 mg/mL
Cross Reactivity QC Testing: Mouse
Format BV480
Product Type BV480 Rat Anti-Mouse CD117
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566081
View Documents Promotion Details
BD
566081
25 μg
Each for $159.00
N/A
BDB566074
View Documents Promotion Details
BD
566074
0.1 mg
Each for $386.00
N/A
Description

Description

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications

Specifications

Flow cytometry
QC Testing: Mouse
BV480 Rat Anti-Mouse CD117
Store undiluted at 4°C
0.2 mg/mL
BV480
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit