Learn More
BD BV480 Rat Anti-Mouse CD45
BD Horizon - BV480 Rat Anti-Mouse CD45 - CD45 is a transmembrane glycoprotein which is expressed at high levels on cell-surface, and its presence distinguishes leukocytes from non-hematopoietic cells. CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction and the CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific.
$192.00 - $296.00
Specifications
Product Type | BV480 Rat Anti-Mouse CD45 |
---|---|
For Use With (Application) | Flow Cytometry |
Concentration | 0.2 mg/mL |
Cross Reactivity | QC Testing: Mouse |
Form | BV480 |
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon BV480 can be excited by the violet laser and detected in the BD Horizon BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Rat Anti-Mouse CD45 | |
0.2 mg/mL | |
BV480 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Flow Cytometry | |
QC Testing: Mouse | |
Store undiluted at 4°C |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.