Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Rat Anti-Mouse IgD

The 11-26c.2a monoclonal antibody specifically binds to mouse immunoglobulin D of all Igh-C haplotypes and it does not react with other immunoglobulin isotypes. BD Horizon™ BV480 Rat Anti-Mouse IgD is expressed on the splenic B-cell surface with high affinity, it does not induce proliferation of splenic B cells in vitro.

Manufacturer:  BD 566199

 View more versions of this product

Catalog No. BDB566199


Add to cart

Description

Description

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications

Specifications

0.2 mg/mL
Flow cytometry
BV480 Rat Anti-Mouse IgD
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
QC Testing: Mouse
BV480
25 μg
Store undiluted at 4°C
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit