Learn More
BD Macro-Vue™ RPR Cards and Components Available on GSA/VA Contract for Federal Government customers only.
For large-scale testing in public health and clinical laboratories
Manufacturer:  BD 275692
 View more versions of this product
Catalog No. 22-349-772
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
RPR Card antigen suspension is a carbon particle cardiolipin antigen1 which detects reagin, an antibody-like substance present in serum or plasma from syphilitic persons, and occasionally in serum or plasma of persons with other acute or chronic conditions. The reagin binds to the test antigen, which consists of cardiolipin-lecithin-coated cholesterol particles, causing macroscopic flocculation.
  • CLIA Complexity: Moderate
  • Nontreponemal test
  • For the serological detection of syphilis as described in A Manual of Tests for Syphilis
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Detects “reagin” present in Serum of syphilitic patients
10,000 Test Kits
Bulk RPR Cards
For large-scale testing in public health and clinical laboratories
10,000 Tests/ Pack