BD Microtainer™ Plastic Capillary Blood Collectors

Designed for collecting, transporting and processing small quantities of blood obtained by skin puncture

Manufacturer: BD 365958

 View more versions of this product

Catalog No. 02-675-182

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Product Type Plasma Tube
    Capacity (Metric) 400μL
    Additive Lithium Heparin w/Gel
    Color Mint Green
    Applications Collecting, transporting and processing small quantities of blood obtained by skin puncture
    Closure Type Color-coded stoppers
    Material Plastic
    Closure Included Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit