Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Microtainer Plastic Capillary Blood Collectors
Designed for collecting, transporting and processing small quantities of blood obtained by skin puncture
Specifications
Applications | Collecting, transporting and processing small quantities of blood obtained by skin puncture |
---|---|
Closure Included | Yes |
Closure Type | Color-coded stoppers |
Material | Plastic |
Catalog Number | Mfr. No. | Additive | Color | Capacity (Metric) | Product Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Additive | Color | Capacity (Metric) | Product Type | Price | Quantity & Availability | ||
02-669-38
|
BD
365973/DEL |
K2 EDTA | Lavender | 500μL | Whole Blood Tube |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
- FloTop collectors (on top of the tube) allow faster and more efficient collection
- Color-coded stoppers simplify sample identification and additive type
- FloCoat process for dispensing EDTA delivers a more uniform distribution throughout the tube, assuring a proper blood-to-additive ratio and faster anti-coagulation
- Volume-level lines on most tubes also help ensure proper blood-to-additive ratio
Specifications
Collecting, transporting and processing small quantities of blood obtained by skin puncture | |
Yes | |
Color-coded stoppers | |
Plastic |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title