Learn More
BD Microtainer™ Plastic Capillary Blood Collectors
Designed for collecting, transporting and processing small quantities of blood obtained by skin puncture
Manufacturer:  BD 365971
 View more versions of this product
Catalog No. 02-668-75
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • FloTop collectors (on top of the tube) allow faster and more efficient collection
  • Color-coded stoppers simplify sample identification and additive type
  • FloCoat™ process for dispensing EDTA delivers a more uniform distribution throughout the tube, assuring a proper blood-to-additive ratio and faster anti-coagulation
  • Volume-level lines on most tubes also help ensure proper blood-to-additive ratio
Specifications
Specifications
Yes
400μL
Color-coded stoppers
Lithium Heparin
Plasma Tube
Collecting, transporting and processing small quantities of blood obtained by skin puncture
Plastic
Green