Learn More
BD Mouse Cerebrum Lysate
Convenient to use and provided in denatured format allowing for immediate loading onto SDS-PAGE gel
Manufacturer:  BD 611455
Catalog No. BDB611455
Add to cart
Includes: Mouse Cerebrum
Description
Description
  • Useful control for western blotting application
  • Rich in endogenously expressed proteins
Specifications
Specifications
Mouse Cerebrum
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
Mouse
500μg
-20°C
1mg/mL