BD Multi-Use Nestable Sharps Collectors

Nestable sharps containers save valuable storage space

Manufacturer: BD 305457

 View more versions of this product

Catalog No. 14-826-68B

  Check Availability
Add to Cart

    Specifications

    Autoclavable Yes
    Capacity (English) 6 gal.
    Material Polypropylene (PP)
    Closure Color Clear
    Capacity (Metric) 22.7L
    Quantity 12 Containers
    Color Red
    For Use With (Application) Suitable for incineration or autoclaving, and general-purpose collectors for safe transport and final disposal
    Closure Type Open Top
    Capacity 6 gal., 22.7L

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit