Learn More
BD Multifit Reusable Luer-Lok™ Syringes
Made of special shockproof glass
Manufacturer:  BD 512441
 View more versions of this product
Catalog No. 14-823-10B
Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Precisely engineered so that every plunger fits every barrel of the same catalog number
  • Calibrated scale fixed into barrel for permanence
  • Molded barrel for friction-free operation
  • Use with PrecisionGlide™, Yale™, and Perfektum™ needles
  • Luer-Lok tips made of chrome-plated brass attached to reinforced heavy glass base
Specifications
Specifications
Syringe
5 mL
PrecisionGlide, Yale and Perfektum Needles
Luer Lock
SDS
Documents
Documents
Product Certifications
Accessories