Learn More
BD Multifit Reusable Luer-Lok™ Syringes
Made of special shockproof glass
Specifications
Material Glass
View More Specs
Products 5
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
14-823-10C
Accessories View Documents
BD
512442
10 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
14-823-10D
Accessories View Documents
BD
512443
20 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
14-823-10B
Accessories View Documents
BD
512441
5 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
14-823-10E
Accessories View Documents
BD
512444
30 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
14-823-10F
Accessories View Documents
BD
512445
50 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
  • Precisely engineered so that every plunger fits every barrel of the same catalog number
  • Calibrated scale fixed into barrel for permanence
  • Molded barrel for friction-free operation
  • Use with PrecisionGlide™, Yale™, and Perfektum™ needles
  • Luer-Lok tips made of chrome-plated brass attached to reinforced heavy glass base
Specifications
Specifications
Glass