Learn More
BD Persist™ Skin Prep Swabs
Fast and easy one-step application
Manufacturer:  BD 386402
 View more versions of this product
Catalog No. 22-265-645
Add to cart
Description
Description
  • 10% PVP-I and 70% alcohol
  • Provides up to seven days of antimicrobial efficacy under an occlusive dressing
  • Lays the foundation for IV catheter start
  • Dries in less than a munute
Specifications
Specifications
Triple Swab
70% Alcohol and 10% Povidone Iodine
Non-sterile
Kills microorganisms immediately on contact
Yes
Safety and Handling
Safety and Handling
ShelfLife : 1825 Day(s)