Learn More
BD PFSK-1 Cell Lysate
Convenient to use and provided in a denatured format allowing for immediate loading onto an SDS-PAGE gel
Manufacturer:  BD 611942
Catalog No. BDB611942
Add to cart
Includes: Rat Primitive Neuroectodermal Tumor, Brain
Description
Description
  • Useful control for western blotting application
  • Can be rich in endogenously expressed proteins
Specifications
Specifications
PFSK-1
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
Rat
500μg
-20°C
1mg/mL