Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosflow Perm Buffer II and Perm Buffer III, BD

For the permeabilization of fixed Peripheral Blood Mononuclear Cells (PBMCs) or lysed whole blood

$213.50

Specifications

Format Liquid
Content And Storage Room Temperature
Quantity 125 mL
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
BDB558052
View Documents
BD Biosciences
558052
Phosflow Perm Buffer II
Each for $213.50
Add to cart
 
BDB558050
View Documents
BD Biosciences
558050
Phosflow Perm Buffer III
Each for $213.50
Add to cart
 
Description

Description

  • Designed for optimal fluorescent staining in flow cytometry
  • Can be used with antibodies for phospho-protein analysis
  • In many cases, may be used simultaneously with phospho-specific antibodies and the staining of cell surface markers (e.g CD markers)

Specifications

Specifications

Liquid
Room Temperature
125 mL
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit