Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD CLIA Waived (Physician) Combo Reader Pack
Includes two packs of B256045 (Flu A + B Test Kit) and one pack of B256055 (Veritor System Reader)
Supplier: BD 256071
Description
Flu A+ B Test Kit 256045
- Unique adaptive-read technology helps reduce false-positive results
- Aids in diagnosis of influenza A and B viral infections
- Incorporates unique Advanced Particle and Adaptive Read technologies
- Sensitive and specific for test
- CLIA Complexity: Waived
- Quantity: 30 pack
Veritor System Reader
- Requires minimal hands-on time
- Easy sample processing: Unitized tube containing correct volume of process reagent facilitates workflow
- Ready in minutes: Test device is ready to insert into reader 10 minutes after sample is added
- Insert and read: Simple one-touch button readies reader for test device insertion
- Results delivered: Once test device is inserted in reader, an objective, digital test result is displayed in 10 seconds
- Cleared for use with nasal swabs and nasopharyngeal (NP) swabs
Specifications
Waived | |
Detection of Flu A + B | |
10 min. |
Combo Reader Pack (Physician) - 256071 | |
Two packs of B256045 (Flu A + B Test Kit) and one B256055 (Veritor System Reader). |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction