Available on GSA/VA Contract for Federal Government customers only. BD PI/RNase Staining Buffer
May be used to analyze cell cycle by flow cytometry or used with antibodies to examine the expression of proteins during the cell cycle
Propidium Iodide (PI) is a fluorescent vital dye that stains DNA and RNA. PI binds to both DNA and RNA, so the latter must be removed by digestion with ribonucleases. The content of DNA as determined by flow cytometry, can reveal useful information about the cell cycle and the proteins involved in the regulation of the cell cycle. Cells in G2 and M phases of the cell cycle have double the DNA content of those in G0 and G1 phases. Cells in S phase have a DNA content lying between these extremes.
$90.00
  • Products 1
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
BDB550825
Available on GSA/VA Contract for Federal Government customers only. 
BD
550825
100mL
Each for $90.00
Check Availability
Add to Cart
Descriptions & Specifications
Related Products