Learn More
BD Plated Media Sabouraud Dextrose Agar
Favorable for the growth of fungi, especially dermatophytes, and slightly inhibitory to contaminating bacteria in clinical specimens
Manufacturer:  BD 299977
Catalog No. 22-349-610
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Specifications
ISO/DIS, 11014
Liquid
Fill 75mL ±5mL, Gel Strength Firm
Plate
Sterile
Light Beige
United States
Sabouraud Dextrose Agar
5 Pk.