Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD PrecisionGlide™ Single-use Needles: Regular Bevel - Regular Wall
Supplier: BD 305129
Description
- PrecisionGlide stainless-steel needles with translucent, color-coded, polypropylene hubs
- Can also be sterilized with ethylene oxide (EtO) in their individual, easy-to-peel blister package
- Regular wall, regular bevel
Order Info
Needles are autoclavable after removal from package.
Specifications
Autoclavable | |
Green | |
Facilitate the injection of high-viscosity fluids | |
Syringe Needle | |
Bevel |
Stainless Steel | |
21 G | |
2 in. | |
51 mm | |
Sterile |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction